Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tricyclohexylphosphonium tetrafluoroborate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38683-0010 38683-0050
TS38683-0010EA 38 USD InStock
TS38683-0010 TS38683-0050
Tricyclohexylphosphonium tetrafluoroborate 99%
Tricyclohexylphosphonium tetrafluoroborate


Formula: C₁₈H₃₄BF₄P
MW: 368.24 g/mol
Melting Pt: 165…169 °C
Storage Temperature: Ambient
MDL Number: MFCD03840580
CAS Number: 58656-04-5
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
Elemental Analysis ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty