Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Dimethylamino)acetaldehyde dimethylacetal 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39091-0010 39091-0050
TS39091-0010EA 30 USD InStock
TS39091-0010 TS39091-0050
(Dimethylamino)acetaldehyde dimethylacetal 97%
(Dimethylamino)acetaldehyde dimethylacetal


Formula: C₆H₁₅NO₂
Boiling Pt: 64…66 °C (12 mmHg)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00010205
CAS Number: 38711-20-5
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4120 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty