Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BAPTA tetrasodium salt hydrate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

40165-0010 40165-0050
TS40165-0010EA 119 USD InStock
TS40165-0010 TS40165-0050
BAPTA tetrasodium salt hydrate 95%
BAPTA tetrasodium salt hydrate


Formula: C₂₂H₂₀N₂Na₄O₁₀·xH₂O
Storage Temperature: Freezer
MDL Number: MFCD02683087
CAS Number: 336624-09-0
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
Titration Complexometric ≥94.0 %
Water ≤12 % (K.F.)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty