Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylene blue hydrate, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41425-0025 41425-1000 41425-5000
TS41425-0025EA 1029 USD InStock
TS41425-0025 TS41425-1000 TS41425-5000
Methylene blue hydrate, pure
Methylene blue hydrate


Formula: C₁₆H₁₈ClN₃S·xH₂O
MW: 319.86 g/mol (anhy)
Melting Pt: 100…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00150006
CAS Number: 122965-43-9
EINECS: 200-515-2
Order Now
Specification Test Results
Appearance Green Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤22 %
Titration with TiCl3 ≥70 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty