Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Naphthoquinone-4-sulphonic acid sodium salt 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41540-0100 41540-0250 41540-1000
TS41540-0100EA 74 USD InStock
TS41540-0100 TS41540-0250 TS41540-1000
1,2-Naphthoquinone-4-sulphonic acid sodium salt 98%
1,2-Naphthoquinone-4-sulphonic acid sodium salt


Formula: C₁₀H₅NaO₅S
Melting Pt: 289 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001700
CAS Number: 521-24-4
EINECS: 208-308-9
Order Now
Specification Test Results
Appearance Orange Powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty