Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodamine 123 99+%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41909-1000 41909-5000
TS41909-1000EA 278 USD InStock
TS41909-1000 TS41909-5000
Rhodamine 123 99+%, pure
Rhodamine 123


Formula: C₂₁H₁₇ClN₂O₃
MW: 380.82 g/mol
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00012664
CAS Number: 62669-70-9
Order Now
Specification Test Results
Appearance Red-brown to brown Powder
Infrared spectrum Conforms
UV spectrophotometry ≥99.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty