Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tetradecyl sulphate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42026-0050 42026-0250
TS42026-0050EA 148 USD InStock
TS42026-0050 TS42026-0250
Sodium tetradecyl sulphate 95%
Sodium tetradecyl sulphate


Formula: C₁₄H₂₉NaO₄S
MW: 316.44
Melting Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00007468
CAS Number: 1191-50-0
Merck Index: 12,08838
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Assay ≥94 %
Additional info Cr2O3: 0.00005 %
Particle size 205 µm (D50) Typical

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty