Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloroisocyanuric acid sodium salt monohydrate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43645-0010 43645-0500 43645-2500
TS43645-0010EA 131 USD InStock
TS43645-0010 TS43645-0500 TS43645-2500
Dichloroisocyanuric acid sodium salt monohydrate 97%
Dichloroisocyanuric acid sodium salt monohydrate


Formula: C₃Cl₂N₃NaO₃.H₂O
MW: 237.96 g/mol
Melting Pt: 240…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00149404
CAS Number: 52671-45-1
EINECS: 220-767-7
UN: 2465
ADR: 5.1 II
Order Now
Specification Test Results
Appearance White Powder or crystals or granules
Infrared spectrum Conforms
Titration Iodometric 96.0 to 104.0 % (on dry substance)
Loss on drying 6 % to 11 % (105°C, 4 h)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty