Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Pyridylazo)resorcinol 97+% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45293-0010 45293-0050
TS45293-0010EA 27 USD InStock
TS45293-0010 TS45293-0050
4-(2-Pyridylazo)resorcinol 97+% ACS
4-(2-Pyridylazo)resorcinol


Formula: C₁₁H₉N₃O₂
MW: 215.21 g/mol
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00044237
CAS Number: 1141-59-9
Order Now
Specification Test Results
Appearance Orange Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.0 %
Suitability (as complexom. indicator) Passes test
Clarity of solution Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty