Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl(3-pyridyl)borane 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45775-0050 45775-0250
TS45775-0050EA 59 USD InStock
TS45775-0050 TS45775-0250
Diethyl(3-pyridyl)borane 97%
Diethyl(3-pyridyl)borane


Formula: C₉H₁₄BN
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00012348
CAS Number: 89878-14-8
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 170 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty