Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Margaric acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

32974-0010 32974-0050 32974-0250
TS32974-0010EA 32 USD InStock
TS32974-0010 TS32974-0050 TS32974-0250
Margaric acid 98%
Margaric acid


Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 364
Melting Pt: 63
Storage Temperature: Ambient
MDL Number: MFCD00002751
CAS Number: 506-12-7
EINECS: 208-027-1
Merck Index: 13,05775
Order Now
Specification Test Results
Appearance White to off-white Glistening crystalline powder or flakes
Infrared spectrum Conforms
Melting point 59 to 63 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty