Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cerotic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

25821-0010 25821-1000
TS25821-0010EA 439 USD InStock
TS25821-0010 TS25821-1000
Cerotic acid 98%
Cerotic acid


Formula: C₂₆H₅₂O₂
Boiling Pt: 250 °C
Melting Pt: 87…89 °C
Density: 0.81 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002811
CAS Number: 506-46-7
EINECS: 208-040-2
Order Now
Specification Test Results
Appearance White Glistening powder
Infrared spectrum Conforms
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty