Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palmitic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12970-0010 12970-0025 12970-0250 12970-2500
TS12970-0010EA 86 USD InStock
TS12970-0010 TS12970-0025 TS12970-0250 TS12970-2500
Palmitic acid 98%
Palmitic acid


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352
Melting Pt: 59…63 °C
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064
Order Now
Specification Test Results
Appearance White to off-white Powder or granules or flakes
Infrared spectrum Conforms
GC ≥97.5 %
Acid value 215 to 233 mg KOH/g
Iodine value ≤1 g I2/100 g
Saponification value 216 to 224 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty