Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% for biochemistry, for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

16762-0010 16762-0051 16762-1000
TS16762-0010EA 157 USD InStock
TS16762-0010 TS16762-0051 TS16762-1000
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% for biochemistry, for analysis
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)


Formula: NH₂C(CH₂OH)₃
Boiling Pt: 219…220 °C (10 mmHg)
Melting Pt: >168.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=b84ca1743f9dfe50eb47c249460ab41a6772ceb2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Heavy metals ≤5 ppm
Sulfated ash ≤0.1 %
UV at 290 nm A: ≤0.2
pH 10.0 to 11.5 (5 % in water)
Titration with HClO4 ≥99.75 % (on dry substance)
Chloride (Cl) ≤5 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤5 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty