Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(+)-Tryptophan 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

16863-0050 16863-0250 16863-1000
TS16863-0050EA 48 USD InStock
TS16863-0050 TS16863-0250 TS16863-1000
D-(+)-Tryptophan 99%
D-(+)-Tryptophan


Formula: C₁₁H₁₂N₂O₂
Melting Pt: 282…285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005647
CAS Number: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Solubility (1 % in water) Clear colorless to light yellow
Specific optical rotation +29.5° to +33.5° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty