Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxynicotinic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15640-0050 15640-0250 15640-1000
TS15640-0050EA 23 USD InStock
TS15640-0050 TS15640-0250 TS15640-1000
6-Hydroxynicotinic acid 99%
6-Hydroxynicotinic acid


Formula: C₆H₅NO₃
Melting Pt: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006277
CAS Number: 5006-66-6
EINECS: 225-682-9
Order Now
Specification Test Results
Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty