Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Serine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

13266-0025 13266-0250 13266-1000 13266-5000
TS13266-0025EA 1499 USD InStock
TS13266-0025 TS13266-0250 TS13266-1000 TS13266-5000
L(+)-Serine 99%
L(+)-Serine


Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
Melting Pt: 222 °C (dec.)
Density: 1.6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals or needles
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, H2O, 430 nm) 1 cm cell
pH 5.0 to 6.5 (2.5 % at 25°C)
Specific optical rotation +14.4° to +16.0° (20°C, 589 nm) (c=10, 2 N HCl)
Titration with HClO4 98.5 to 101.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids ≤0.5 % (TLC)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty