Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Hydroxy-L(-)-proline 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12178-0100 12178-0250 12178-1000 12178-5000
TS12178-0100EA 50 USD InStock
TS12178-0100 TS12178-0250 TS12178-1000 TS12178-5000
trans-4-Hydroxy-L(-)-proline 99+%
trans-4-Hydroxy-L(-)-proline


Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 296
Melting Pt: 206 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, H2O, 430 nm) 1 cm cell
Specific optical rotation -74° to -77° (20°C, 589 nm) (c=5, H2O)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids ≤0.5 % (TLC)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty