Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Isonipecotic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12262-0250 12262-1000 12262-5000
TS12262-0250EA 52 USD InStock
TS12262-0250 TS12262-1000 TS12262-5000
(±)-Isonipecotic acid 99%
(±)-Isonipecotic acid


Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 264
Melting Pt: 157
Storage Temperature: Ambient
MDL Number: MFCD00006004
CAS Number: 498-94-2
EINECS: 207-872-3
Merck Index: 13,05208
Order Now
Specification Test Results
Appearance White to pink-beige Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (120°C, 4 hrs)
Solubility (10 % in water) Clear to slightly hazy
Titration with HClO4 ≥98.5 % (On dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty