Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carboxymethyl cellulose sodium salt, M.W. ∼ 250000 (DS=0.9)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

33262-0010 33262-1000
TS33262-0010EA 163 USD InStock
TS33262-0010 TS33262-1000
Carboxymethyl cellulose sodium salt, M.W. ∼ 250000 (DS=0.9)
Carboxymethyl cellulose sodium salt


Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Assay ≥99.5 %
Loss on drying ≤10 % (As packed) (3 to 5 g, 105°C, 2 h)
Heavy metals ≤10 ppm
Degree of substitution 0.80 to 0.95
Viscosity 1500 to 3100 mPa.s (2 % at 25°C) (Brookfield)
pH 6.5 to 8 (1 % solution)
Sodium chloride (NaCl) ≤0.25 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤3 ppm
Mercury (Hg) ≤1 ppm
Impurity ≤0.4 % (Sodium glycolate)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty