Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Cinchonidine 98.5-101% (dry basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11033-0250 11033-1000
TS11033-0250EA 67 USD InStock
TS11033-0250 TS11033-1000
(-)-Cinchonidine 98.5-101% (dry basis)
(-)-Cinchonidine


Formula: C₁₉H₂₂N₂O
Boiling Pt: 465 °C (1013 hPa)
Melting Pt: 200…203 °C
MDL Number: MFCD00006783
CAS Number: 485-71-2
EINECS: 207-622-3
Merck Index: 13,02307
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point ≥200 °C
Loss on drying ≤1 % (105 °C)
Specific optical rotation -105° to -115° (20 °C, 589 nm) (c=1, EtOH) (on dried substance)
Titration with HClO4 98.5 to 101.0 % (On dried substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty