Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CAPS (3-(cyclohexylamino)propanesulphonic acid) 99% for biochemistry

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17262-0010 17262-1000 17262-2500
TS17262-0010EA 588 USD InStock
TS17262-0010 TS17262-1000 TS17262-2500
CAPS (3-(cyclohexylamino)propanesulphonic acid) 99% for biochemistry
CAPS (3-(cyclohexylamino)propanesulphonic acid)


Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
Melting Pt: 324 (dec.)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
UV at 280 nm A: ≤0.02
Titration with NaOH ≥98.5 % (On dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty