Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium tetrathionate dihydrate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46192-0250 46192-1000
TS46192-0250EA 167 USD InStock
TS46192-0250 TS46192-1000
Sodium tetrathionate dihydrate
Sodium tetrathionate dihydrate


Formula: Na₂O₆S₄·2H₂O
MW: 306.27
Storage Temperature: Refrigerator
MDL Number: MFCD00150868
CAS Number: 13721-29-4
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Solubility colorless to light yellow
Titration after Ion exchange ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty