Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Threonine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34697-0050 34697-0250 34697-1000
TS34697-0050EA 46 USD InStock
TS34697-0050 TS34697-0250 TS34697-1000
D-Threonine 98%
D-Threonine


Formula: C₄H₉NO₃
Melting Pt: 274 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064269
CAS Number: 632-20-2
EINECS: 211-171-8
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
UV transmittance ≥98 % (c=5, H2O, 430 nm) 1 cm cell
Specific optical rotation +27° to +29° (20°C, 589 nm) (c=6, water)
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids (TLC) Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty