Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-α-Phenylglycine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

21799-0250 21799-1000
TS21799-0250EA 32 USD InStock
TS21799-0250 TS21799-1000
L(+)-α-Phenylglycine 99%
L(+)-α-Phenylglycine


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064403
CAS Number: 2935-35-5
EINECS: 220-909-8
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 h)
Specific optical rotation +154° to +160° (20°C, 589 nm) (c=1, 1 M HCl)
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty