Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Shikimic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

13270-0050 13270-0010
TS13270-0050EA 427 USD InStock
TS13270-0050 TS13270-0010
(-)-Shikimic acid 98%
(-)-Shikimic acid


Formula: C₇H₁₀O₅
Melting Pt: 185…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00066278
CAS Number: 138-59-0
EINECS: 205-334-2
Order Now
Specification Test Results
Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 183 to 191 °C
Specific optical rotation -170° to -185° (20 °C, 589 nm) (C=2, H2O)
Titration with NaOH 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty