Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12010-1000 12010-5000
TS12010-1000EA 59 USD InStock
TS12010-1000 TS12010-5000
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 98%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)


Formula: C₃H₈ClNO₂
MW: 125.56 g/mol
Melting Pt: 173…177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012870
CAS Number: 5680-79-5
EINECS: 227-139-1
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
GC ≥95 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty