Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bacitracin zinc salt, potency min. 60 Units/mg

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22611-0010 22611-0050
TS22611-0010EA 52 USD InStock
TS22611-0010 TS22611-0050
Bacitracin zinc salt, potency min. 60 Units/mg
Bacitracin zinc salt


Formula: C₆₆H₁₀₁N₁₇O₁₆SZn
Melting Pt: 250 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00130598
CAS Number: 1405-89-6
EINECS: 215-787-8
Order Now
Specification Test Results
Appearance White to yellow to beige to light grey Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 %
Potency ≥60 U/mg (On dried substance)
pH 6.0 to 7.5 (10 % solution)
Zinc (Zn) 4.0 to 6.0 % (on dried substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty