Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Bromoadenosine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15578-0010 15578-0050
TS15578-0010EA 126 USD InStock
TS15578-0010 TS15578-0050
8-Bromoadenosine 98%
8-Bromoadenosine


Formula: C₁₀H₁₂BrN₅O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00005733
CAS Number: 2946-39-6
EINECS: 220-959-0
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -53° to -56° (20°C, 589 nm) (c=2.5, DMSO)
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty