Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

99% for biochemistry

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

17260-1000 17260-5000
TS17260-1000EA 126 USD InStock
TS17260-1000 TS17260-5000
99% for biochemistry
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid)


Formula: C₉H₂₀N₂O₄S
MDL Number: MFCD00006160
CAS Number: 16052-06-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
UV at 260 nm A: ≤0.1
pH 5 to 6 (10 % w/v at 20°C)
Titration with NaOH ≥98.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty