Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(-)-Isoascorbic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22727-5000 22727-0025
TS22727-5000EA 58 USD InStock
TS22727-5000 TS22727-0025
D-(-)-Isoascorbic acid 98%
D(-)-Isoascorbic acid


Formula: C₆H₈O₆
MW: 176.12 g/mol
Melting Pt: 167…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00005378
CAS Number: 89-65-6
EINECS: 201-928-0
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 164 to 172 °C
Titration Iodimetric ≥97.5 %
Loss on drying ≤0.4 %
Sulfated ash ≤0.3 %
Specific optical rotation -16.5° to -18° (25 °C, 589 nm) (c=10, water)
Lead (Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty