Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(+)-Arabitol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

30287-0050 30287-0250 30287-1000
TS30287-0050EA 46 USD InStock
TS30287-0050 TS30287-0250 TS30287-1000
D-(+)-Arabitol 99%
D(+)-Arabitol


Formula: C₅H₁₂O₅
Melting Pt: 97…105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004709
CAS Number: 488-82-4
EINECS: 207-686-2
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 105 °C
HPLC ≥98.5 % (ELSD)
Specific optical rotation +10.5° to +13.5° (20 °C, 589 nm) (c=2, 2 % borax)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty