Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-piperidineacetic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43750-0010 43750-0050
TS43750-0010EA 196 USD InStock
TS43750-0010 TS43750-0050
1-Boc-4-piperidineacetic acid 97%
1-Boc-4-piperidineacetic acid


Formula: C₁₂H₂₁NO₄
Melting Pt: 99…100.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00800239
CAS Number: 157688-46-5
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 102 °C
Titration with NaOH 96 to 104 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty