Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Choline hydroxide 45% (w/w) in aqueous solution stabilised, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

38672-0025 38672-0100 38672-1000 38672-5000
TS38672-0025EA 872 USD InStock
TS38672-0025 TS38672-0100 TS38672-1000 TS38672-5000
Choline hydroxide 45% (w/w) in aqueous solution stabilised, pure
Choline hydroxide


Formula: C₅H₁₅NO₂
Density: 1.07 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00002831
CAS Number: 123-41-1
UN: 3267
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to tan Viscous solution
Stabilizer about 0.2 % ethylenediamine
Titration with HClO4 44.0 to 48.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty