Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N(α)-Acetyl-L-tryptophan 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

45165-0050 45165-0250 45165-1000
TS45165-0050EA 37 USD InStock
TS45165-0050 TS45165-0250 TS45165-1000
N(α)-Acetyl-L-tryptophan 97%
N(α)-Acetyl-L-tryptophan


Formula: C₁₃H₁₄N₂O₃
Melting Pt: 186…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065976
CAS Number: 1218-34-4
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 %
Loss on drying ≤0.3 %
Specific optical rotation +23° to +28° (20°C, 589 nm) (c=1, EtOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty