Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-L-valine 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

27571-0050 27571-0250 27571-1000
TS27571-0050EA 40 USD InStock
TS27571-0050 TS27571-0250 TS27571-1000
N-Boc-L-valine 99+%
N-Boc-L-valine


Formula: C₁₀H₁₉NO₄
MW: 217.3 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065605
CAS Number: 13734-41-3
EINECS: 237-307-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
HPLC ≥99.0 % (ELSD)
Specific optical rotation -5.3° to -7.3° (20 °C, 589 nm) (c=1, CH3COOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty