Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-α-methyl-4-biphenylacetic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

45466-0010 45466-0050 45466-0250
TS45466-0010EA 39 USD InStock
TS45466-0010 TS45466-0050 TS45466-0250
2-Fluoro-α-methyl-4-biphenylacetic acid 98%
2-Fluoro-α-methyl-4-biphenylacetic acid


Formula: C₁₅H₁₃FO₂
MW: 244.265 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00079303
CAS Number: 5104-49-4
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 110 to 116 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty