Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluorothymidine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45291-0010 45291-2500
TS45291-0010EA 551 USD InStock
TS45291-0010 TS45291-2500
Trifluorothymidine 98%
Trifluorothymidine


Formula: C₁₀H₁₁F₃N₂O₅
MW: 296.2 g/mol
Melting Pt: 190…193 °C
Storage Temperature: Freezer
MDL Number: MFCD00006534
CAS Number: 70-00-8
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty