Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Proline methyl ester hydrochloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30300-0010 30300-0050 30300-0250
TS30300-0010EA 27 USD InStock
TS30300-0010 TS30300-0050 TS30300-0250
L-Proline methyl ester hydrochloride 98%
L-Proline methyl ester hydrochloride


Formula: C₆H₁₁NO₂·HCl
MW: 165.6 g/mol
Melting Pt: 71 °C
Storage Temperature: Freezer
MDL Number: MFCD00012708
CAS Number: 2133-40-6
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Water ≤0.5 % (K.F.)
Specific optical rotation -31° to -35° (20°C, 589 nm) (c=1, H2O)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty