Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',3'-O-Isopropylideneadenosine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12272-0050 12272-0250
TS12272-0050EA 66 USD InStock
TS12272-0050 TS12272-0250
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine


Formula: C₁₃H₁₇N₅O₄
Melting Pt: 221…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00005756
CAS Number: 362-75-4
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV spectrophotometry molar absorptivity 15400 +/-4%, on dry substance
Ratio of Absorbances ≤0.01 (A290/A260)
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty