Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

46525-0010 46525-0050
TS46525-0010EA 62 USD InStock
TS46525-0010 TS46525-0050
3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one
3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one


Formula: C₂₅H₂₈N₆O
Melting Pt: 180…182 °C
Storage Temperature: Freezer
MDL Number: MFCD00864464
CAS Number: 138402-11-6
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 180 to 184 °C
Loss on drying ≤2 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty