Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46233-0010 46233-0050
TS46233-0010EA 133 USD InStock
TS46233-0010 TS46233-0050
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride


Formula: C₂₂H₂₄ClN₃O₄
MW: 429.901 g/mol
Melting Pt: 215…227 °C
Storage Temperature: Ambient
MDL Number: MFCD07781272
CAS Number: 183319-69-9
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Solubility (1% in DMSO) Clear solution
HPLC ≥97.5 %
Residue after ignition ≤0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty