Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Butenoic acid 90% tech. unstabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15883-0250 15883-1000
TS15883-0250EA 99 USD InStock
TS15883-0250 TS15883-1000
3-Butenoic acid 90% tech. unstabilised
3-Butenoic acid


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163 °C (760 mmHg)
Melting Pt: –39 °C
Density: 1.01 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002782
CAS Number: 625-38-7
UN: 3265
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥88 %
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Heavy metals (as Pb) ≤0.001 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty