Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-DL-tryptophan 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12797-0010 12797-0050 12797-2500
TS12797-0010EA 300 USD InStock
TS12797-0010 TS12797-0050 TS12797-2500
5-Methyl-DL-tryptophan 98%
5-Methyl-DL-tryptophan


Formula: C₁₂H₁₄N₂O₂
MW: 218.26 g/mol
Melting Pt: 280…282 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005652
CAS Number: 951-55-3
Order Now
Specification Test Results
Appearance Off-white Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Sulfated ash ≤0.5 %
Appearance of solution (5% soln. in 1 N NaOH) clear, pale yellowish
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty