Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anastrozole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

46097-0010 46097-2500
TS46097-0010EA 551 USD InStock
TS46097-0010 TS46097-2500
Anastrozole 98%
Anastrozole


Formula: C₁₇H₁₉N₅
Melting Pt: 81…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00866298
CAS Number: 120511-73-1
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 82 to 86 °C
Water ≤0.3 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty