Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azelastine hydrochloride

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46508-0010 46508-2500
TS46508-0010EA 216 USD InStock
TS46508-0010 TS46508-2500
Azelastine hydrochloride
Azelastine hydrochloride


Formula: C₂₂H₂₄ClN₃O
Melting Pt: 225…229 °C
Storage Temperature: Freezer
MDL Number: MFCD00242783
CAS Number: 79307-93-0
Order Now
Specification Test Results
Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 224 to 228 °C
HPLC ≥97.5 %
Titration Argentimetric 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty