Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Alanine methyl ester hydrochloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

43532-0050 43532-0250
TS43532-0050EA 49 USD InStock
TS43532-0050 TS43532-0250
D-Alanine methyl ester hydrochloride 98%
D-Alanine methyl ester hydrochloride


Formula: C₄H₉NO₂
Melting Pt: 60.4…62.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00066141
CAS Number: 14316-06-4
Order Now
Specification Test Results
Appearance White Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
Specific optical rotation -10° to -15° (20°C, 589 nm) c=2, methanol

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty