Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Isoserine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

27967-0010 27967-2500
TS27967-0010EA 141 USD InStock
TS27967-0010 TS27967-2500
DL-Isoserine 98%
DL-Isoserine


Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00008138
CAS Number: 565-71-9
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty