Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Levamisole hydrochloride 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18787-0100 18787-0500
TS18787-0100EA 81 USD InStock
TS18787-0100 TS18787-0500
Levamisole hydrochloride 99+%
Levamisole hydrochloride


Formula: C₁₁H₁₂N₂S·HCl
Melting Pt: 226…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00012675
CAS Number: 16595-80-5
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 4 hrs)
Specific optical rotation -121° to -128° (20°C, 589 nm) (c=5, H2O) (on dried substance)
Titration with NaOH ≥99.0 % (On dried substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty