Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lovastatin 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45883-0050 45883-0250 45883-1000
TS45883-0050EA 98 USD InStock
TS45883-0050 TS45883-0250 TS45883-1000
Lovastatin 98%
Lovastatin


Formula: C₂₄H₃₆O₅
MW: 404.55 g/mol
Melting Pt: 175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072164
CAS Number: 75330-75-5
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty